missing translation for 'onlineSavingsMsg'
Learn More

(-)-Riboflavin, TRC

Product Code. 30428244
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30428244 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30428244 Supplier Toronto Research Chemicals Supplier No. TRCR41499525G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Vitamin B2 (Riboflavin)
CAS 83-88-5
Purity Grade Notes HPLC
Molecular Formula C17 H20 N4 O6
Quantity 25 g
Synonym Riboflavin, Riboflavine (7CI), Benzo[g]pteridine','riboflavin deriv., (-)-Riboflavin, 1-Deoxy-1-(3,4-dihydro-7,8-dimethyl-2,4-dioxobenzo[g]pteridin-10(2H)-yl)-D-ribitol, 6,7-Dimethyl-9-D-ribitylisoalloxazine, 6,7-Dimethyl-9-ribitylisoalloxazine, Beflavin, Beflavine, Benzo[g]pteridine-2,4(3H,10H)-dione','7,8-dimethyl-10-(D-ribo-2,3,4,5-tetrahydroxypentyl)-, C.I. 50900, C.I. Food Yellow 15, D-Ribitol','1-deoxy-1-(3,4-dihydro-7,8-dimethyl-2,4-dioxobenzo[g]pteridin-10(2H)-yl)-, E 101, E 101 (dye), Flavaxin, Flavin BB, Flaxain, Food Yellow 15, Hyre, Lactobene, Lactoflavin, Lactoflavine, NCI 0033298, NSC 33298, Ribipca, Ribocrisina, Riboderm, Ribosyn, Ribotone, Ribovel, Russupteridine yellow III, San Yellow B, Vitaflavine, Vitamin B2, Vitamin G, Vitasan B2
InChI Formula InChI=1S/C17H20N4O6/c1-7-3-9-10(4-8(7)2)21(5-11(23)14(25)12(24)6-22)15-13(18-9)16(26)20-17(27)19-15/h3-4,11-12,14,22-25H,5-6H2,1-2H3,(H,20,26,27)/t11-,12+,14-/m0/s1
SMILES Cc1cc2N=C3C(=O)NC(=O)N=C3N(C[C@H](O)[C@H](O)[C@H](O)CO)c2cc1C
IUPAC Name 7,8-dimethyl-10-[(2S,3S,4R)-2,3,4,5-tetrahydroxypentyl]benzo[g]pteridine-2,4-dione
Molecular Weight (g/mol) 376.36
Formula Weight 376.1383
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.