missing translation for 'onlineSavingsMsg'
Learn More

Riboflavin Phosphate Sodium Salt (~70%), TRC

Product Code. 17959924
Change view
Click to view available options
Quantity:
100 mg
1 g
10 g
Unit Size:
100mg
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17959924 100 mg 100mg
17907464 1 g 1g
17909564 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17959924 Supplier Toronto Research Chemicals Supplier No. TRCR415000100MG
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 130-40-5
Molecular Formula C17 H20 N4 O9 P . Na
Molecular Weight (g/mol) 478.33
Synonym Riboflavin sodium phosphate, Riboflavin 5'-(sodium hydrogen phosphate), Riboflavin 5'-(dihydrogen phosphate), sodium salt (1:1), Riboflavin 5'-(dihydrogen phosphate), monosodium salt (9CI), Riboflavine 5'-(dihydrogen phosphate), monosodium salt (6CI,8CI), Riboflavine, 5'-phosphate, sodium salt (7CI), Benzo[g]pteridine, riboflavin 5'-(dihydrogen phosphate) deriv., AMN, Alloxazine mononucleotide sodium salt, Coflavinase, Cytoflav, Flamotide, Hyryl, Ribo, Riboflavin 5'-phosphate ester monosodium salt, Riboflavin 5'-phosphate sodium, Riboflavin monophosphate monosodium salt, Riboflavin phosphate sodium, Riboflavin-5-phosphoric acid monosodium salt, Riboflavine monosodium 5'-phosphate, Riboflavine phosphate sodium, Riboflavine sodium phosphate, Riboflavine-5-phosphate sodium, Sodium FMN, Sodium riboflavin phosphate, Sodium riboflavin-5'-phosphate
IUPAC Name sodium;[(2R,3S,4S)-5-(7,8-dimethyl-2,4-dioxobenzo[g]pteridin-10-yl)-2,3,4-trihydroxypentyl] hydrogen phosphate
SMILES [Na+].Cc1cc2N=C3C(=O)NC(=O)N=C3N(C[C@H](O)[C@H](O)[C@H](O)COP(=O)(O)[O-])c2cc1C

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C17H21N4O9P.Na/c1-7-3-9-10(4-8(7)2)21(15-13(18-9)16(25)20-17(26)19-15)5-11(22)14(24)12(23)6-30-31(27,28)29;/h3-4,11-12,14,22-24H,5-6H2,1-2H3,(H,20,25,26)(H2,27,28,29);/q;+1/p-1/t11-,12+,14-;/m0./s1
Formula Weight 478.0866
Chemical Name or Material Riboflavin-5'-Phosphate sodium salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.