missing translation for 'onlineSavingsMsg'
Learn More

Riboflavin phosphate sodium, MedChemExpress

Product Code. 16482226
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16482226 500 mg 500mg
16478536 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16482226 Supplier Medchem Express Supplier No. HYB0964
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Riboflavin phosphate sodium (FMN-Na) is a derivative of Riboflavin (vitamin B2) which is an essential nutrient for animals. Riboflavin phosphate sodium can be used for the research of progressive keratoconus, corneal ectasia and irregular astigmatism. Riboflavine phosphate sodium is a very effective NAD+-recycling agent.

Riboflavin phosphate sodium (FMN-Na) is a derivative of Riboflavin (vitamin B2) which is an essential nutrient for animals. Riboflavin phosphate sodium can be used for the research of progressive keratoconus, corneal ectasia and irregular astigmatism. Riboflavine phosphate sodium is a very effective NAD+-recycling agent.

Specifications

Chemical Name or Material Riboflavin phosphate sodium
CAS 130-40-5
Color Orange
Purity Grade Notes Research
Molecular Formula C17H20N4NaO9P
Quantity 500 mg
Synonym FMN-Na Riboflavin 5'-phosphate sodium Vitamin B2 Phosphate Sodium Salt
Solubility Information H2O : 20.83 mg/mL (43.55 mM; ultrasonic and warming and heat to 60°C) ∣DMSO : 1 mg/mL (2.09 mM; Need ultrasonic)
SMILES CC1=CC2=C(C=C1C)N=C(C(N2C[C@H](O)[C@H](O)[C@H](O)COP(O)(O[Na])=O)=N3)C(NC3=O)=O
Molecular Weight (g/mol) 478.33
Formula Weight 478.33
Percent Purity 92.37%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.