missing translation for 'onlineSavingsMsg'
Learn More

Riboflavin (Fine, Orange-Yellow Crystals/Electrophoresis), Fisher BioReagents

Product Code. 10796644
Change view
Click to view available options
Quantity:
50 g
Packaging:
Glass Bottle
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10796644 50 g Glass Bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10796644 Supplier Fisher BioReagents Supplier No. BP16750
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 83-88-5 | C17H20N4O6 | 376.37 g/mol

Glass Bottle; 50g

Riboflavin is a photoinitiator of acrylamide/bis-acrylamide polymerization.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Riboflavin
CAS 83-88-5
Melting Point 290°C
Color Yellow to Orange
pH 5.5 to 7.2
Assay Percent Range ≥95 %
Packaging Glass Bottle
Molecular Formula C17H20N4O6
MDL Number MFCD00005022
Quantity 50 g
Merck Index 15, 8325
Synonym riboflavin, vitamin b2, lactoflavin, riboflavine, vitamin g, lactoflavine, --riboflavin, beflavin, beflavine, flavaxin
InChI Key AUNGANRZJHBGPY-QTZZOOGMNA-N
SMILES CC1=C(C)C=C2N(C[C@H](O)[C@H](O)[C@H](O)CO)C3=NC(=O)NC(=O)C3=NC2=C1
IUPAC Name 7,8-dimethyl-10-[(2S,3S,4R)-2,3,4,5-tetrahydroxypentyl]benzo[g]pteridine-2,4-dione
Molecular Weight (g/mol) 376.37
PubChem CID 71310809
Formula Weight 374.04
Percent Purity ≥95%
Grade Electrophoresis
Physical Form Solid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.