missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Riboflavin, 98%

Product Code. 10135783 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 10135783

Brand: Thermo Scientific Chemicals 132355000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 83-88-5 | C17H20N4O6 | 376.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 83-88-5
Molecular Formula C17H20N4O6
Molecular Weight (g/mol) 376.37
MDL Number MFCD00005022
InChI Key AUNGANRZJHBGPY-QTZZOOGMNA-N
Synonym riboflavin, vitamin b2, lactoflavin, riboflavine, vitamin g, lactoflavine, --riboflavin, beflavin, beflavine, flavaxin
PubChem CID 71310809
SMILES CC1=C(C)C=C2N(C[C@H](O)[C@H](O)[C@H](O)CO)C3=NC(=O)NC(=O)C3=NC2=C1

Specifications

Absorbance 0.31 to 0.33 (A373/A267), 0.36 to 0.39 (A444/A267)
Melting Point 290.0°C
Color Yellow to Orange
Loss on Drying 1.5% max. (105°C)
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 500 g
Merck Index 15, 8325
Specific Rotation Condition − 125.00 (20.00°C c=0.5,0.05 M NaOH)
Specific Rotation -125
Solubility Information Solubility in water: 0.07g/L (20°C). Other solubilities: very soluble in diluted alkalies,,slightly soluble in ethanol and nacl solutions,,insoluble in ethers,chloroform,acetone,benzene,and lipid solvents
Spectrophotometry 97.5% min. (lambda max. 444nm) (A (1%/1 cm) 328,on dried substance)
IUPAC Name 7,8-dimethyl-10-(2,3,4,5-tetrahydroxypentyl)-2H,3H,4H,10H-benzo[g]pteridine-2,4-dione
Formula Weight 376.36
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Riboflavin
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.