missing translation for 'onlineSavingsMsg'
Learn More

Riboflavin-5'-phosphate sodium salt dihydrate

Product Code. 11491918
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11491918 10 g 10g
11401928 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11491918 Supplier Thermo Scientific Alfa Aesar Supplier No. A14545.09
  • ONLINE EXCLUSIVE PRICE
    £28.00
    £25.65 / 10g
    Save £2.35
    8% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6184-17-4 | C17H24N4NaO11P | 514.36 g/mol

Riboflavin 5?-monophosphate (FMN) is used as a prosthetic group by oxidoreductase (one and two electron transfer) enzymes such as NADH dehydrogenase, nitric oxide synthase, and nitrilotriacetate monooxygenase. Riboflavin 5?-monophosphate is used as a substrate to study the specificity and kinetics of FMN phosphohydrolases and to study molecular sensing based gene regulation by riboswitches. It is a photoinitiator of polymerization for PAGE gels.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Riboflavin 5′-monophosphate (FMN) is used as a prosthetic group by oxidoreductase (one and two electron transfer) enzymes such as NADH dehydrogenase, nitric oxide synthase, and nitrilotriacetate monooxygenase. Riboflavin 5′-monophosphate is used as a substrate to study the specificity and kinetics of FMN phosphohydrolases and to study molecular sensing based gene regulation by riboswitches. It is a photoinitiator of polymerization for PAGE gels.

Solubility
Soluble in water 50 g/L @ 20°C.

Notes
Store in refrigerator. Protect from heat. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6184-17-4
Molecular Formula C17H24N4NaO11P
Molecular Weight (g/mol) 514.36
MDL Number MFCD00150993
InChI Key LQVGUKOCMOKKJU-FTNKLFHDNA-M
Synonym riboflavin 5'-monophosphate sodium salt dihydrate
PubChem CID 131856174
IUPAC Name [(2R,3S,4S)-5-(7,8-dimethyl-2,4-dioxobenzo[g]pteridin-10-yl)-2,3,4-trihydroxypentyl] dihydrogen phosphate;sodium;dihydrate
SMILES O.O.[Na+].CC1=C(C)C=C2N(C[C@H](O)[C@H](O)[C@H](O)COP(O)([O-])=O)C3=NC(=O)NC(=O)C3=NC2=C1

Spezifikation

Melting Point >300°C
Quantity 10 g
Beilstein 4106529
Merck Index 14,8201
Solubility Information Soluble in water 50g/L @ 20°C.
Formula Weight 514.36 (478.34 Anhydrous)
Chemical Name or Material Riboflavin-5'-phosphate sodium salt dihydrate

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.