missing translation for 'onlineSavingsMsg'
Learn More

Ribocil-C, MedChemExpress

Product Code. 16487778
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16487778 10 mg 10mg
16437626 100 mg 100mg
16430248 5 mg 5mg
16403747 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16487778 Supplier Medchem Express Supplier No. HY19488A
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Ribocil-C is a highly selective inhibitor of bacterial riboflavin riboswitches.

Ribocil-C is a highly selective inhibitor of bacterial riboflavin riboswitches.

Specifications

Chemical Name or Material Ribocil-C
CAS 1825355-56-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C21H21N7OS
Quantity 10 mg
Solubility Information DMSO : 110 mg/mL (262.22 mM; Need ultrasonic)
SMILES O=C1C=C(C2=CC=CS2)NC([C@H]3CCCN(CC4=CN(C5=NC=CC=N5)C=N4)C3)=N1
Molecular Weight (g/mol) 419.5
Formula Weight 419.5
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.