missing translation for 'onlineSavingsMsg'
Learn More

Ribociclib, MedChemExpress

Product Code. 16469367
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16469367 5 mg 5mg
16442897 50 mg 50mg
16426828 10 mg 10mg
16426936 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16469367 Supplier Medchem Express Supplier No. HY15777
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Ribociclib (LEE01) is a highly specific CDK4/6 inhibitor with IC50 values of 10 nM and 39 nM, respectively, and is over 1,000-fold less potent against the cyclin B/CDK1 complex.

Ribociclib (LEE01) is a highly specific CDK4/6 inhibitor with IC50 values of 10 nM and 39 nM, respectively, and is over 1,000-fold less potent against the cyclin B/CDK1 complex.

Specifications

Chemical Name or Material Ribociclib
CAS 1211441-98-3
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C23H30N8O
Quantity 5 mg
Synonym LEE011
Solubility Information DMSO : 20 mg/mL (46.03 mM; Need ultrasonic)
SMILES O=C(N(C)C)C(N1C2CCCC2)=CC(C1=N3)=CN=C3NC(N=C4)=CC=C4N5CCNCC5
Molecular Weight (g/mol) 434.54
Formula Weight 434.54
Percent Purity 99.52%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.