missing translation for 'onlineSavingsMsg'
Learn More

Ribociclib hydrochloride, MedChemExpress

Product Code. p-200097745
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16479367

Brand: Medchem Express HY15777A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Ribociclib hydrochloride (LEE011 hydrochloride) is a highly specific CDK4/6 inhibitor with IC50 values of 10 nM and 39 nM, respectively, and is over 1,000-fold less potent against the cyclin B/CDK1 complex.

Ribociclib hydrochloride (LEE011 hydrochloride) is a highly specific CDK4/6 inhibitor with IC50 values of 10 nM and 39 nM, respectively, and is over 1,000-fold less potent against the cyclin B/CDK1 complex.

Specifications

Chemical Name or Material Ribociclib hydrochloride
CAS 1211443-80-9
Color Yellow
Purity Grade Notes Research
Molecular Formula C23H31ClN8O
Quantity 5 mg
Synonym LEE011 hydrochloride
Solubility Information H2O : 10 mg/mL (21.23 mM; Need ultrasonic) ∣DMSO : 7.69 mg/mL (16.33 mM; Need ultrasonic)
SMILES O=C(N(C)C)C(N1C2CCCC2)=CC(C1=N3)=CN=C3NC(N=C4)=CC=C4N5CCNCC5.Cl
Molecular Weight (g/mol) 471.0
Formula Weight 471.0
Percent Purity 99.2%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.