missing translation for 'onlineSavingsMsg'
Learn More

Ribitol, MedChemExpress

Product Code. 16411526
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16411526 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16411526 Supplier Medchem Express Supplier No. HY100582

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Ribitol is a crystalline pentose alcohol formed by the reduction of ribose. Enhancing the flux of D-glucose to the pentose phosphate pathway in Saccharomyces cerevisiae for the production of D-ribose and ribitol.

Ribitol is a crystalline pentose alcohol formed by the reduction of ribose. Enhancing the flux of D-glucose to the pentose phosphate pathway in Saccharomyces cerevisiae for the production of D-ribose and ribitol.

Specifications

Chemical Name or Material Ribitol
Physical Form Powder
CAS 488-81-3
Color White
Quantity 500 mg
Purity Grade Notes Research
Molecular Formula C5H12O5
Synonym Adonitol Adonite
Solubility Information H2O : 33.33 mg/mL (219.06 mM; Need ultrasonic) ∣DMSO : 1.1 mg/mL (7.23 mM; Need ultrasonic)
SMILES OC[C@@H](O)[C@@H](O)[C@@H](O)CO
Molecular Weight (g/mol) 152.15
Formula Weight 152.15
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.