missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RIBC2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA551861
This item is not returnable.
View return policy
Description
Immunogen sequence: DKIMCILENR KKRDRKNLCR AINDFQQSFQ KPETRREFDL SDPLALKKDL PARQSDNDVR NTISGMQKFM GEDLNFHERK KFQEEQNREW SLQQQREWKN ARAEQKCAEA LYTETRLQFD ETAKHLQKLE STTRKAVCAS VKDF Highest antigen sequence identity to the following orthologs: Mouse - 72%, Rat - 76%.
RIBC2 (RIB43A Domain With Coiled-Coils 2) is a Protein Coding gene. An important paralog of this gene is RIBC1.
Specifications
| RIBC2 | |
| Polyclonal | |
| Unconjugated | |
| RIBC2 | |
| 4930579A10Rik; C22orf11; RGD1309033; RIB43A domain with coiled-coils 2; RIB43A-like with coiled-coils protein 2; RIBC2; testis-specific ribbon protein; Trib | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 26150 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9H4K1 | |
| RIBC2 | |
| Recombinant protein corresponding to Human RIBC2. Recombinant protein control fragment (Product #RP-91515). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction