missing translation for 'onlineSavingsMsg'
Learn More

Ribavirin, MedChemExpress

Product Code. 16471416
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16471416 200 mg 200mg
16448306 100 mg 100mg
16432136 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16471416 Supplier Medchem Express Supplier No. HYB0434
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Ribavirin (ICN-1229) is an antiviral agent against a broad spectrum of viruses including HCV, HIVl, and RSV. Ribavirin also has anti-orthopoxvirus and anti-variola activities.

Ribavirin (ICN-1229) is an antiviral agent against a broad spectrum of viruses including HCV, HIVl, and RSV. Ribavirin also has anti-orthopoxvirus and anti-variola activities.

Specifications

Chemical Name or Material Ribavirin
Physical Form Powder
CAS 36791-04-5
Color White
Quantity 200 mg
Purity Grade Notes Research
Molecular Formula C8H12N4O5
For Use With (Application) COVID-19-anti-virus
Synonym ICN-1229
Solubility Information H2O : 100 mg/mL (409.50 mM; Need ultrasonic) ∣DMSO : 100 mg/mL (409.50 mM; Need ultrasonic)
SMILES O[C@H]1[C@H](N2N=C(C(N)=O)N=C2)O[C@H](CO)[C@H]1O
Molecular Weight (g/mol) 244.2
Formula Weight 244.2
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.