missing translation for 'onlineSavingsMsg'
Learn More

RIAM/APBB1IP Antibody [PE], Novus Biologicals™

Product Code. 30736532 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30736532 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30736532 Supplier Novus Biologicals Supplier No. AF7279P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

RIAM/APBB1IP Polyclonal antibody specifically detects RIAM/APBB1IP in Human,Mouse samples. It is validated for Western Blot,Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen RIAM/APBB1IP
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate PE
Formulation PBS
Gene Alias amyloid beta (A4) precursor protein-binding, family B, member 1 interactingprotein, amyloid beta A4 precursor protein-binding family B member 1-interacting protein, APBB1-interacting protein 1, INAG1, PREL1, PREL-1, proline rich EVH1 ligand 1, Proline-rich EVH1 ligand 1, Proline-rich protein 73, Rap1-GTP-interacting adapter molecule, Rap1-interacting adaptor molecule, RARP1, Retinoic acid-responsive proline-rich protein 1, RIAMRARP-1
Host Species Sheep
Immunogen E. coli-derived recombinant human RIAM/APBB1IP, Gly2-Ser89, Accession # Q7Z5R6
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54518
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.