missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RIAM/APBB1IP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£328.00 - £513.00
Specifications
| Antigen | RIAM/APBB1IP |
|---|---|
| Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18675775
|
Novus Biologicals
NBP2-39009-25ul |
25 μL |
£328.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18197877
|
Novus Biologicals
NBP2-39009 |
0.1 mL |
£513.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RIAM/APBB1IP Polyclonal specifically detects RIAM/APBB1IP in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| RIAM/APBB1IP | |
| Polyclonal | |
| Rabbit | |
| Human | |
| Q7Z5R6 | |
| 54518 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: KTLYDNYQRAVAKAGLASRWTNLGTVNAAAPAQPSTGPKTGTTQPNGQIPQATHSVSAVLQEAQRHAETSKDKKPAL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| amyloid beta (A4) precursor protein-binding, family B, member 1 interactingprotein, amyloid beta A4 precursor protein-binding family B member 1-interacting protein, APBB1-interacting protein 1, INAG1, PREL1, PREL-1, proline rich EVH1 ligand 1, Proline-rich EVH1 ligand 1, Proline-rich protein 73, Rap1-GTP-interacting adapter molecule, Rap1-interacting adaptor molecule, RARP1, Retinoic acid-responsive proline-rich protein 1, RIAMRARP-1 | |
| APBB1IP | |
| IgG | |
| Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title