missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RIAM/APBB1IP Antibody (7E7A3) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52417-0.025mg
This item is not returnable.
View return policy
Description
RIAM/APBB1IP Monoclonal specifically detects RIAM/APBB1IP in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.
Specifications
| RIAM/APBB1IP | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, CyTOF-ready | |
| amyloid beta (A4) precursor protein-binding, family B, member 1 interactingprotein, amyloid beta A4 precursor protein-binding family B member 1-interacting protein, APBB1-interacting protein 1, INAG1, PREL1, PREL-1, proline rich EVH1 ligand 1, Proline-rich EVH1 ligand 1, Proline-rich protein 73, Rap1-GTP-interacting adapter molecule, Rap1-interacting adaptor molecule, RARP1, Retinoic acid-responsive proline-rich protein 1, RIAMRARP-1 | |
| Mouse | |
| 73.2 kDa | |
| 0.025 mg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, CyTOF | |
| 7E7A3 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| APBB1IP | |
| Purified recombinant fragment of human APBB1IP (AA: 1-151) expressed in E. Coli. | |
| Protein A or G purified | |
| RUO | |
| 54518 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction