missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RHPN1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103248
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total RHPN1.
Has no enzymatic activity. May serve as a target for Rho, and interact with some cytoskeletal component upon Rho binding or relay a Rho signal to other molecules.
Specifications
| RHPN1 | |
| Polyclonal | |
| Unconjugated | |
| RHPN1 | |
| BB023497; Grbp; GTP-rho binding protein 1; GTP-Rho-binding protein 1; KIAA1929; mKIAA1929; ODF5; outer dense fiber of sperm tails 5; RHOPHILIN; rhophilin 1; rhophilin Rho GTPase binding protein 1; rhophilin, Rho GTPase binding protein 1; rhophilin-1; RHPN; RHPN1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 114822 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8TCX5 | |
| RHPN1 | |
| A synthesized peptide derived from human RHPN1(Accession Q8TCX5), corresponding to amino acid residues 651-665. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction