missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RHOXF2 Mouse anti-Human, Clone: PCRP-RHOXF2-1D7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-07401-100ug
This item is not returnable.
View return policy
Description
RHOXF2 Monoclonal specifically detects RHOXF2 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Protein Array.
Specifications
| RHOXF2 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 1 to 2 μg/million cells, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL, Protein Array | |
| CT107, Paired-like homeobox protein PEPP-2, PEPP subfamily gene 2, PEPP-2, PEPP2cancer/testis antigen 107, rhox homeobox family member 2, Rhox homeobox family, member 2, Testis homeobox gene 1, THG1homeobox protein from AL590526 | |
| Recombinant full-length human RHOXF2 protein | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunofluorescence, Peptide Array | |
| PCRP-RHOXF2-1D7 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 84528 | |
| Store at 4°C. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction