missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RHOXF2 Antibody (PCRP-RHOXF2-1D7), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08529MFV450
This item is not returnable.
View return policy
Description
RHOXF2 Monoclonal antibody specifically detects RHOXF2 in Human samples. It is validated for Flow Cytometry
Specifications
| RHOXF2 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 84528 | |
| Store at 4C in the dark. | |
| IgG2b |
| Flow Cytometry | |
| PCRP-RHOXF2-1D7 | |
| Flow Cytometry | |
| CT107, Paired-like homeobox protein PEPP-2, PEPP subfamily gene 2, PEPP-2, PEPP2cancer/testis antigen 107, rhox homeobox family member 2, Rhox homeobox family, member 2, Testis homeobox gene 1, THG1homeobox protein from AL590526 | |
| Recombinant full-length human RHOXF2 protein (Uniprot: Q9BQY4 ) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido