missing translation for 'onlineSavingsMsg'
Learn More

RHOXF2 Antibody (PCRP-RHOXF2-1D7), Alexa Fluor™ 750, Novus Biologicals™

Product Code. 18589110 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18589110 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18589110 Supplier Novus Biologicals Supplier No. NBP308529AF750

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

RHOXF2 Monoclonal antibody specifically detects RHOXF2 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Protein Array
TRUSTED_SUSTAINABILITY

Specifications

Antigen RHOXF2
Applications Flow Cytometry, Immunofluorescence, Protein Array
Classification Monoclonal
Clone PCRP-RHOXF2-1D7
Conjugate Alexa Fluor 750
Formulation 50mM Sodium Borate
Gene Alias CT107, Paired-like homeobox protein PEPP-2, PEPP subfamily gene 2, PEPP-2, PEPP2cancer/testis antigen 107, rhox homeobox family member 2, Rhox homeobox family, member 2, Testis homeobox gene 1, THG1homeobox protein from AL590526
Host Species Mouse
Immunogen Recombinant full-length human RHOXF2 protein (Uniprot: Q9BQY4 )
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84528
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.