missing translation for 'onlineSavingsMsg'
Learn More

RHOXF1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Product Code. 18336374 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18336374 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18336374 Supplier Novus Biologicals Supplier No. NBP310458100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RHOXF1 Polyclonal specifically detects RHOXF1 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RHOXF1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS buffer, 2% sucrose
Gene Alias MGC119030, OTEXMGC119033, Ovary-, testis- and epididymis-expressed gene protein, Paired-like homeobox protein PEPP-1, PEPP subfamily gene 1, PEPP1paired-like homeobox protein OTEX, rhox homeobox family member 1, Rhox homeobox family, member 1
Host Species Rabbit
Immunogen The immunogen is a synthetic peptide directed towards the C terminal region of human RHOXF1 (NP_644811). Peptide sequence VPTRRELAENLGVTEDKVRVWFKNKRARCRRHQRELMLANELRADPDDCV
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 158800
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.