missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RHOXF1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£362.00
Specifications
| Antigen | RHOXF1 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
RHOXF1 Polyclonal specifically detects RHOXF1 in Human samples. It is validated for Western Blot.Specifications
| RHOXF1 | |
| Polyclonal | |
| Rabbit | |
| Signal Transduction | |
| Q8NHV9 | |
| 158800 | |
| Synthetic peptides corresponding to RHOXF1 (Rhox homeobox family, member 1) The peptide sequence was selected from the N terminal of RHOXF1. Peptide sequence PEGGVNHENGMNRDGGMIPEGGGGNQEPRQQPQPPPEEPAQAAMEGPQPE. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| MGC119030, OTEXMGC119033, Ovary-, testis- and epididymis-expressed gene protein, Paired-like homeobox protein PEPP-1, PEPP subfamily gene 1, PEPP1paired-like homeobox protein OTEX, rhox homeobox family member 1, Rhox homeobox family, member 1 | |
| RHOXF1 | |
| IgG | |
| 20 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title