missing translation for 'onlineSavingsMsg'
Learn More

Rhox6 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Product Code. 18303986 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18303986 25 μg 25µL
18319694 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18303986 Supplier Novus Biologicals Supplier No. NBP31040925UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Rhox6 Polyclonal specifically detects Rhox6 in Mouse samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Spezifikation

Antigen Rhox6
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS buffer, 2% sucrose
Gene Alias Homeobox protein PSX, mCG_17933, MCG17933, Protein Rhox6, Psx1, reproductive homeobox 6, Reproductive homeobox on X chromosome 6, Rhox6 reproductive homeobox 6
Host Species Rabbit
Immunogen The immunogen is a synthetic peptide corresponding to a region of Mouse (NP_032981). Peptide sequence GRHAGDGASSSEDDSILEEGGENIDQQPPQQEAASPDSIRNPHVLNRLAQ
Purification Method Affinity purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19202
Target Species Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.