missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RhoV Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£222.00 - £514.00
Specifications
| Antigen | RhoV |
|---|---|
| Dilution | Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18684019
|
Novus Biologicals
NBP2-49082 |
0.1 mL |
£514.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18677455
|
Novus Biologicals
NBP2-49082-25ul |
25 μL |
£222.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RhoV Polyclonal antibody specifically detects RhoV in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| RhoV | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| PBS (pH 7.2), 40% Glycerol | |
| 171177 | |
| IgG | |
| Immunogen affinity purified |
| Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| ARHV, CDC42-like GTPase 2, CHP, GTP-binding protein-like 2, ras homolog gene family, member V, Rho GTPase-like protein ARHV, rho-related GTP-binding protein RhoV, Wnt-1 regulated Cdc42 homolog 2, Wnt-1 responsive Cdc42 homolog 2, WRCH1-related GTPase, WRCH-2, WRCH2Chp | |
| This antibody was developed against a recombinant protein corresponding to amino acids: KEVFDSAILSAIEHKARLEKKLNAKGVRTLSRCRWKKFFCFV | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title