missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Rhotekin Monoclonal Antibody (OTI1A4)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA526038
This item is not returnable.
View return policy
Description
Rhotekin Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
This gene encodes a scaffold protein that interacts with GTP-bound Rho proteins. Binding of this protein inhibits the GTPase activity of Rho proteins. This protein may interfere with the conversion of active, GTP-bound Rho to the inactive GDP-bound form by RhoGAP. Rho proteins regulate many important cellular processes, including cytokinesis, transcription, smooth muscle contraction, cell growth and transformation. Dysregulation of the Rho signal transduction pathway has been implicated in many forms of cancer. Alternative splicing results in multiple transcript variants encoding different isoforms.
Specifications
| Rhotekin | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9BST9 | |
| RTKN | |
| Full length human recombinant protein of RTKN produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| OTI1A4 | |
| Unconjugated | |
| RTKN | |
| hypothetical protein LOC539691; RGD:727874}; rhotekin; RTKN; rtkn {ECO:0000312; RTKN1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 6242 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction