missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RHOT2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
£247.00 - £396.00
Specifications
| Antigen | RHOT2 |
|---|---|
| Dilution | Western Blot 0.04-0.4 ug/ml, Simple Western 1:20, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:50-1:200 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18402581
|
Novus Biologicals
NBP1-88982-25ul |
25 μL |
£247.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18293636
|
Novus Biologicals
NBP1-88982 |
0.1 mL |
£396.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RHOT2 Polyclonal specifically detects RHOT2 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| RHOT2 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| ARHT2, C16orf39, chromosome 16 open reading frame 39, EC 3.6.5, EC 3.6.5.-, hMiro-2, MIRO-2mitochondrial Rho 2, mitochondrial Rho GTPase 2, Ras homolog gene family member T2, ras homolog gene family, member T2, RASL | |
| RHOT2 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot 0.04-0.4 ug/ml, Simple Western 1:20, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:50-1:200 | |
| Polyclonal | |
| Rabbit | |
| Signal Transduction | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 89941 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:RSCLGHLGYLGYPTLCEQDQAHAITVTREKRLDQEKGQTQRSVLLCKVVGACGVGKSAFLQAFLGRGLGHQDTREQPPGYAIDTVQVNGQEKYLILC | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title