missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Rhosin hydrochloride
GREENER_CHOICE

Product Code. 18764709
Click to view available options
Quantity:
50 mg
1 x 10 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18764709

Brand: Tocris Bioscience™ 5003/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rho GTPase inhibitor

Rhosin hydrochloride is a Rho GTPase inhibitor (Kd = 354 nM). Inhibits binding of RhoA to guanine nucleotide exchange factors (GEFs) including LARG, DBL, LBC, p115 RhoGEF and PDZ RhoGEF. Has no effect on the binding of Rac1 or Cdc42 to their respective GEFs. Suppresses RhoA/C-mediated breast cancer cell migration and invasion in vitro. Also induces neurite outgrowth in synergy with NGF.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 50 mg
Product Type Rhosin hydrochloride
Purity >98%
Molecular Weight (g/mol) 394.86
Inhibitors Rho GTPase Inhibitors

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.