missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RhoJ Antibody (1E4), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00057381-M01
This item is not returnable.
View return policy
Description
RhoJ Monoclonal antibody specifically detects RhoJ in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Functional Assay
Specifications
| RhoJ | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| ARHJTC10B, FLJ14445, ras homolog gene family, member J, RASL7B, Ras-like protein family member 7B, RAS-like, family 7, member B, RHOI, rho-related GTP-binding protein RhoJ, Tc10-like GTP-binding protein, TC10-like Rho GTPase, TCLMGC34777 | |
| RHOJ (AAH62575, 1 a.a. ∽ 214 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MNCKEGTDSSCGCRGNDEKKMLKCVVVGDGAVGKTCLLMSYANDAFPEEYVPTVFDHYAVTVTVGGKQHLLGLYDTAGQEDYNQLRPLSYPNTDVFLICFSVVNPASYHNVQEEWVPELKDCMPHVPYVLIGTQIDLRDDPKTLARLLYMKEKPLTYEHGVKLAKAIGAQCYLECSALTQKGLKAVFDEAILTIFHPKKKKKRCSEGHSCCSII | |
| 0.1 mg | |
| Signal Transduction | |
| 57381 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG1 λ |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Functional Assay | |
| 1E4 | |
| Western Blot 1:500, ELISA 1:100-1:2000, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1:10-1:500, Functional | |
| AAH62575 | |
| Mouse | |
| IgG purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction