missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RhoGDI Antibody (2G3) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£184.00 - £453.00
Specifications
| Antigen | RhoGDI |
|---|---|
| Clone | 2G3 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, CyTOF |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18697495
|
Novus Biologicals
NBP2-37472-0.025ml |
0.025 mL |
£184.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18195949
|
Novus Biologicals
NBP2-37472 |
0.1 mL |
£453.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RhoGDI Monoclonal specifically detects RhoGDI in Human, Mouse, Primate, Monkey samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, CyTOF-ready.Specifications
| RhoGDI | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, CyTOF | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Primate, Monkey | |
| GDIA1, MGC117248, Rho GDI 1, Rho GDP dissociation inhibitor (GDI) alpha, rho GDP-dissociation inhibitor 1, RHOGDI, Rho-GDI alpha, RHOGDI-1 | |
| ARHGDIA | |
| IgG1 | |
| Affinity Purified |
| 2G3 | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, CyTOF-ready | |
| Monoclonal | |
| Mouse | |
| Signal Transduction | |
| PBS with 0.05% Sodium Azide | |
| 396 | |
| Purified recombinant fragment of human ARHGDIA (AA: FULL(1-204)) expressed in E. Coli. | |
| Primary | |
| 26 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title