missing translation for 'onlineSavingsMsg'
Learn More

RhoGAP Antibody (OTI9H2) [PerCP], Novus Biologicals™

Product Code. 30687828 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30687828 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30687828 Supplier Novus Biologicals Supplier No. NBP344958PCP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

RhoGAP Monoclonal antibody specifically detects RhoGAP in Human,Mouse,Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen RhoGAP
Applications Western Blot
Classification Monoclonal
Clone OTI9H2
Conjugate PerCP
Formulation PBS
Gene Alias growth factor independent 2, p100 RasGAP-associated p105 protein, p105 RhoGAP, p190-BGFI2, p190BRhoGAP, Rho GTPase activating protein 5, rho GTPase-activating protein 5, RHOGAP5, Rho-type GTPase-activating protein 5
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 948-1261 of human ARHGAP5 (NP_001164) produced in E.coli.
Purification Method >95%
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, GPCR, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 394
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.