missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RhoG Polyclonal Antibody

RhoG Polyclonal Antibody
Brand: Thermo Scientific PA5114494
This item is not returnable.
View return policy
Description
RhoG Polyclonal Antibody for Western Blot, IHC (P)
Specifications
| RhoG | |
| Polyclonal | |
| Unconjugated | |
| RHOG | |
| 2810426G09Rik; aplysia ras-related homolog G (RhoG); Arhg; ras homolog family member G; ras homolog G (RhoG); ras homolog gene family, member G; ras homolog gene family, member G (rho G); Rho G; Rhog; rho-related GTP-binding protein RhoG; Sid 10750; Sid10750 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 308875, 391, 56212 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| P84095, P84096 | |
| RHOG | |
| RHOG antibody was raised against a 15 amino acid peptide near the amino terminus of human RHOG. The immunogen is located within amino acids 40 - 90 of RHOG. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction