missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RhoG Antibody (PSH02-70), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-32902
This item is not returnable.
View return policy
Description
RhoG Monoclonal antibody specifically detects RhoG in Human,Monkey samples. It is validated for Flow Cytometry,Western Blot
Specifications
| RhoG | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:2000, Flow Cytometry 1:1000 | |
| ARHGMGC125835, MGC125836, ras homolog gene family, member G (rho G), RhoG, rho-related GTP-binding protein RhoG | |
| Synthetic peptide within human RhoG aa 1-50/ 191. (Uniprot: P84095) | |
| 100 μL | |
| Cell Cycle and Replication | |
| 391 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Flow Cytometry, Western Blot | |
| PSH02-70 | |
| Unconjugated | |
| PBS (pH7.4), 0.1% BSA and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Monkey | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction