missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RhoF Antibody (1D4B6) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52553
This item is not returnable.
View return policy
Description
RhoF Monoclonal specifically detects RhoF in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.
Specifications
| RhoF | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, CyTOF-ready | |
| FLJ20247, ras homolog gene family, member F (in filopodia), Rho family GTPase Rif, Rho in filopodia, rho-related GTP-binding protein RhoF, RIFARHF | |
| Mouse | |
| 23.6 kDa | |
| 0.1 mg | |
| Signal Transduction | |
| 54509 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2a |
| Western Blot, Flow Cytometry, ELISA, CyTOF | |
| 1D4B6 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| RHOF | |
| Purified recombinant fragment of human RHOF (AA: 1-84) expressed in E. Coli. | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction