missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rhodopsin Rabbit anti-Mouse, Rat, Clone: 5N3G9, Novus Biologicals™
Rabbit Monoclonal Antibody
£167.00 - £395.00
Specifications
| Antigen | Rhodopsin |
|---|---|
| Clone | 5N3G9 |
| Dilution | Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18309016
|
Novus Biologicals
NBP3-16655-20UL |
20 μg |
£167.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18325523
|
Novus Biologicals
NBP3-16655-100UL |
100 μg |
£395.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Rhodopsin Monoclonal antibody specifically detects Rhodopsin in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| Rhodopsin | |
| Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse, Rat | |
| CSNBAD1, MGC138309, OPN2MGC138311, opsin 2, rod pigment, opsin-2, retinitis pigmentosa 4, autosomal dominant, rhodopsin, RP4 | |
| A synthetic peptide corresponding to a sequence within amino acids 249-348 of human Rhodopsin (P08100). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| 5N3G9 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| GPCR, Signal Transduction, Vision | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 6010 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title