Learn More
Rhodopsin Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 30438-1-AP-150UL
Description
The Rhodopsin antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human Rhodopsin. This antibody recognizes human, mouse, rat antigen. The Rhodopsin antibody has been validated for the following applications: IHC, ELISA analysis.
Specifications
| Rhodopsin | |
| 500 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat | |
| 150 kDa | |
| IgG |
| Immunohistochemistry, ELISA | |
| CSNBAD1, OPN2, Opsin 2, RHO, rhodopsin, RP4 | |
| 150 μL | |
| 6010 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |