missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rhodopsin, Rabbit anti-Human,Mouse,Rat,Amphibian,Bovine,Zebrafish, Clone: Rho 1D4, Novus Biologicals™
Rabbit Monoclonal Antibody
£226.00 - £473.00
Specifications
| Antigen | Rhodopsin |
|---|---|
| Clone | Rho 1D4 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Radioimmunoassay |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Radioimmune Assays (RIA) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18389410
|
Novus Biologicals
NBP2-81059-0.025MG |
0.025 mg |
£226.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18248128
|
Novus Biologicals
NBP2-81059-0.2MG |
0.2 mg |
£473.00
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Rhodopsin Monoclonal specifically detects Rhodopsin in Human, Mouse, Rat, Amphibian, Bovine, Zebrafish samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Radioimmunoassay.Specifications
| Rhodopsin | |
| 1 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Radioimmune Assays (RIA) | |
| Unconjugated | |
| Rabbit | |
| GPCR, Phospho Specific, Signal Transduction, Vision | |
| PBS | |
| 6010 | |
| Bleached bovine rod outer segment (ROS). | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Rho 1D4 | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Radioimmunoassay | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat, Amphibian, Bovine, Zebrafish | |
| CSNBAD1, MGC138309, OPN2MGC138311, opsin 2, rod pigment, opsin-2, retinitis pigmentosa 4, autosomal dominant, rhodopsin, RP4 | |
| RHO | |
| IgG κ | |
| Protein A purified | |
| The antibody binds to the C-terminus of rhodopsin on the cytoplasmic side of disk membranes. The epitope has the following amino acid sequence: 'TETSQVAPA'. A limitation of the 1D4 tag, however, is that it has to be placed at the C-terminus of a protein. This is because the Rho1D4 monoclonal antibody requires a free carboxylate group for high affinity binding. Amidation of the carboxyl group lowers its immunoreactivity to the Rho1D4 antibody by over 100 fold. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title