Learn More
Epredia™ Lab Vision™ Rhodopsin (Opsin) Ab-1 Mouse Monoclonal Antibody, 200μg/mL, biotinylated, BSA and azide
Provide accurate, reproducible results with the Epredia™ Rhodopsin (Opsin) Ab-1, Mouse Monoclonal Antibody.
£104.04 - £601.00
Specifications
| Antigen | Rhodopsin (Opsin) Ab-1 |
|---|---|
| Clone | RET-P1 |
| Applications | Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Host Species | Mouse |
| Product Code | Brand | Quantity | Form | Concentration | Price | Quantity & Availability | |||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Form | Concentration | Price | Quantity & Availability | |||
|
12609336
|
Epredia™
MS-1233-B0 |
100 μL | Concentrated | 200μg/mL | |||||
|
12619336
|
Epredia™
MS-1233-P0 |
100 μL | Concentrated | 200μg/mL | |||||
|
12639336
|
Epredia™
MS-1233-P1ABX |
100 μL | Concentrated | 1mg/mL | |||||
|
12649336
|
Epredia™
MS-1233-PABX |
200 μL | Concentrated | 1mg/mL | |||||
|
12689326
|
Epredia™
MS-1233-B1 |
500 μL | Concentrated | 200μg/mL | |||||
|
12629336
|
Epredia™
MS-1233-P1 |
500 μL | Concentrated | 200μg/mL |
£342.00 £320.83 / 500µL Save £21.17 6% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
|
12679326
|
Epredia™
MS-1233-B |
1 mL | Concentrated | 200μg/mL | |||||
|
12699326
|
Epredia™
MS-1233-P |
1 mL | Concentrated | 200μg/mL | |||||
|
12679336
|
Epredia™
MS-1233-R7 |
7 mL | Prediluted | Prediluted | |||||
Description
Rhodopsin is a glycoprotein which is a visual pigment in mammals, birds, amphibians, and reptiles. The rhodopsin protein is present on the photoreceptor cells in the retina.Host Species: Mouse
Clone: RET-P1
Isotype: IgG1
Species Reactivity: Human, Mouse and Rat. Others not known.
Epitope: Not determined
Immunogen: Membrane preparation from adult rat retina
Molecular Weight: 39kDa
Positive Control: Brain or retina
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
Note:
USA: RUO; Int'l: RUO
Specifications
| Rhodopsin (Opsin) Ab-1 | |
| Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| Mouse | |
| Cancer and Tumor Biology | |
| Membrane preparation from adult rat retina | |
| Primary |
| RET-P1 | |
| Monoclonal | |
| RUO | |
| Human, Mouse, Rat | |
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.