missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Rhodopsin Monoclonal antibody specifically detects Rhodopsin in Human, Mouse, Rat, Amphibian, Bovine, Zebrafish samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Radioimmune Assays
Specifications
Specifications
| Antigen | Rhodopsin |
| Applications | Western Blot |
| Classification | Monoclonal |
| Dilution | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Radioimmunoassay |
| Formulation | 50mM Sodium Borate |
| Gene Alias | CSNBAD1, MGC138309, OPN2MGC138311, opsin 2, rod pigment, opsin-2, retinitis pigmentosa 4, autosomal dominant, rhodopsin, RP4 |
| Gene ID (Entrez) | 6010 |
| Target Species | Human, Mouse |
| Form | Purified |
| Isotype | IgG κ |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?