missing translation for 'onlineSavingsMsg'
Learn More

Rhodopsin Antibody (Rho 1D4), CoraFluor™ 1, Novus Biologicals™

Product Code. 30150500
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30150500 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30150500 Supplier Novus Biologicals (Bio-Techne) Supplier No. NBP281059CL1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Rhodopsin Monoclonal antibody specifically detects Rhodopsin in Amphibian,Bovine,Human,Mouse,Rat,Zebrafish samples. It is validated for ELISA,Immunohistochemistry,Immunoprecipitation,Western Blot,Radioimmune Assays,Immunocytochemistry/Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rhodopsin
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot, Radioimmune Assays (RIA)
Classification Monoclonal
Clone Rho 1D4
Conjugate CoraFluor 1
Formulation PBS
Gene Alias CSNBAD1, MGC138309, OPN2MGC138311, opsin 2, rod pigment, opsin-2, retinitis pigmentosa 4, autosomal dominant, rhodopsin, RP4
Host Species Rabbit
Immunogen Bleached bovine rod outer segment (ROS).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR, Phospho Specific, Signal Transduction, Vision
Primary or Secondary Primary
Gene ID (Entrez) 6010
Target Species Amphibian, Bovine, Human, Mouse, Rat
Content And Storage Store at 4°C in the dark. Do not freeze.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.