missing translation for 'onlineSavingsMsg'
Learn More

Rhodizonic acid disodium salt, 98%

Product Code. 11413167
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11413167 5 g 5g
11423167 25 g 25g
11433167 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11413167 Supplier Thermo Scientific Alfa Aesar Supplier No. A16126.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 523-21-7 | C6Na2O6 | 214.04 g/mol

Determining sulfate radical by barium chloride titration with rhodizonic acid disodium salt test paper as indicator has been established. Staining of thin sections for quick identification of quartz, potash feldspar and plagioclase is usually done with sodium hexanitrocobaltate (III) and rhodizonic acid disodium salt. N-allyl-N?-(sodium p-phenyl-sulfonate)thiourea is used for eliminating the interference of Ag +?Au 3+?Hg 2+in the identification of Sr 2+ using Rhodizonic acid disodium salt.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Determining sulfate radical by barium chloride titration with rhodizonic acid disodium salt test paper as indicator has been established. Staining of thin sections for quick identification of quartz, potash feldspar and plagioclase is usually done with sodium hexanitrocobaltate (III) and rhodizonic acid disodium salt. N-allyl-N′-(sodium p-phenyl-sulfonate)thiourea is used for eliminating the interference of Ag +?Au 3+?Hg 2+in the identification of Sr 2+ using Rhodizonic acid disodium salt.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 523-21-7
Molecular Formula C6Na2O6
Molecular Weight (g/mol) 214.04
MDL Number MFCD00001650
InChI Key SXWPCWBFJXDMAI-UHFFFAOYSA-L
Synonym sodium rhodizonate, sodium 3,4,5,6-tetraoxocyclohex-1-ene-1,2-bis olate, rhodizonic acid disodium salt, unii-9qf0329l3g, rhodizonic acid sodium salt, rhodizonic acid, disodium salt, disodium 3,4,5,6-tetraoxocyclohexene-1,2-diolate, 3,4,5,6-tetraoxocyclohexene-1,2-diol disodium salt, disodium tetraoxocyclohex-1-ene-1,2-bis olate, dipotassium tetraoxocyclohex-1-ene-1,2-bis olate
PubChem CID 68225
IUPAC Name disodium;3,4,5,6-tetraoxocyclohexene-1,2-diolate
SMILES [Na+].[Na+].[O-]C1=C([O-])C(=O)C(=O)C(=O)C1=O

Specifications

Melting Point >300°C
Odor Odorless
Quantity 5 g
Beilstein 5181379
Merck Index 14,8670
Solubility Information Soluble in water.
Formula Weight 214.04
Percent Purity 98%
Chemical Name or Material Rhodizonic acid disodium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.