missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rhodium Standard For ICP/MS, SPEX CertiPrep™
Click to view available options
Quantity:
30 mL
Packaging:
Triple-leached LDPE bottle
Description
Includes
NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
Rh | 10μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
CAS | 7440-16-6 |
Molecular Formula | Rh |
MDL Number | MFCD00011201 MFCD03458392 |
InChI Key | MHOVAHRLVXNVSD-UHFFFAOYSA-N |
SMILES | [Rh] |
IUPAC Name | rhodium |
Molecular Weight (g/mol) | 102.91 |
PubChem CID | 23948 |
ChEBI | CHEBI:33359 |
Packaging | Triple-leached LDPE bottle |
Show More |
Product intended for laboratory use only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'