missing translation for 'onlineSavingsMsg'
Learn More

Rhodium(III) sulfate tetrahydrate, Premion™, 99.99% (metals basis), Rh 35.9% min, Thermo Scientific Chemicals

Product Code. 11379417
Click to view available options
Quantity:
0.25 g
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Rhodium(III) sulfate is used as plating reagents and additives for noble metal.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15274-78-9
Molecular Formula H8O16Rh2S3
Molecular Weight (g/mol) 566.04
MDL Number MFCD00798544
InChI Key JJITWKYISWZJBI-UHFFFAOYSA-H
Synonym rhodium iii sulfate tetrahydrate, 3so4.2rh.4h2o, rhodium iii sulfate tetrahydrate, premion, dirhodium 3+ tetrahydrate trisulfate
PubChem CID 91886287
IUPAC Name rhodium(3+);trisulfate;tetrahydrate
SMILES O.O.O.O.[Rh+3].[Rh+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

Color Brown
Physical Form Crystalline
Quantity 0.25 g
Linear Formula Rh2(SO4)3·4H2O
UN Number UN3261
Solubility Information Slightly soluble in water.
Formula Weight 566.05 (493.99 Anhydrous)
Percent Purity 99.99%
Grade Premion™
Concentration or Composition (by Analyte or Components) Rh 35.9% min.
Odor Odorless
Assay (metals basis)
Chemical Name or Material Rhodium(III) sulfate tetrahydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'