missing translation for 'onlineSavingsMsg'
Learn More

Rhodium(III) sulfate tetrahydrate, Premion™, 99.99% (metals basis), Rh 35.9% min

Product Code. 11389417
Change view
Click to view available options
Quantity:
0.25 g
1 g
5 g
Unit Size:
0.25g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11389417 1 g 1g
11379417 0.25 g 0.25g
11399417 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11389417 Supplier Thermo Scientific Alfa Aesar Supplier No. 041016.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 15274-78-9 | H8O16Rh2S3 | 566.04 g/mol

Rhodium(III) sulfate is used as plating reagents and additives for noble metal.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15274-78-9
Molecular Formula H8O16Rh2S3
Molecular Weight (g/mol) 566.04
MDL Number MFCD00798544
InChI Key JJITWKYISWZJBI-UHFFFAOYSA-H
Synonym rhodium iii sulfate tetrahydrate, 3so4.2rh.4h2o, rhodium iii sulfate tetrahydrate, premion, dirhodium 3+ tetrahydrate trisulfate
PubChem CID 91886287
IUPAC Name rhodium(3+);trisulfate;tetrahydrate
SMILES O.O.O.O.[Rh+3].[Rh+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

Color Brown
Physical Form Crystalline
Quantity 1 g
Linear Formula Rh2(SO4)3·4H2O
UN Number UN3261
Solubility Information Slightly soluble in water.
Formula Weight 566.05 (493.99 Anhydrous)
Percent Purity 99.99%
Grade Premion
Concentration or Composition (by Analyte or Components) Rh 35.9% min.
Odor Odorless
Assay (metals basis)
Chemical Name or Material Rhodium(III) sulfate tetrahydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.