missing translation for 'onlineSavingsMsg'
Learn More

Rhodium(III) sulfate, bright plating solution, Rh 5% (cont. Rh), Thermo Scientific™

Product Code. 11430310
Change view
Click to view available options
Quantity:
0.5 g
Unit Size:
0.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11430310 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11430310 Supplier Thermo Scientific Alfa Aesar Supplier No. 012635.NJ
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10489-46-0
Molecular Formula O12Rh2S3
Molecular Weight (g/mol) 493.98
MDL Number MFCD00016309
InChI Key YWFDDXXMOPZFFM-UHFFFAOYSA-H
Synonym rhodium iii sulfate solution, rhodium 3+ trisulfate, dirhodium trisulphate, rhodium sulfate, rhodium sulfate solution, acmc-1bntj, sulfuric acid, rhodium 3+ salt 3:2, rhodium iii sulfate, solution, dirhodium 3+ trisulfate, sulfuric acid,rhodium 3+ salt 3:2
PubChem CID 159290
IUPAC Name rhodium(3+);trisulfate
SMILES [Rh+3].[Rh+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

Color Red to Orange
Linear Formula Rh2(SO4)3
UN Number UN3264
Concentration Rh 5% (cont. Rh)
Density 1.4 g/mL
Solubility Information Miscible with water,ethanol and with a 3:1 ethanol/water mixture.
Formula Weight 493.99
Odor Acidic
Quantity 0.5 g
Physical Form Liquid
Chemical Name or Material Rhodium(III) sulfate, Bright plating solution
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.