missing translation for 'onlineSavingsMsg'
Learn More

Rhodium(III) oxide pentahydrate, Premion™, 99.99% (metals basis), Rh 59% min

Product Code. p-7049072
Click to view available options
Quantity:
0.25 g
1 g
5 g
Unit Size:
0.25g
1g
5g
This item is not returnable. View return policy

Product Code. 11367497

Brand: Thermo Scientific Alfa Aesar 012667.01

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 39373-27-8 | H10O8Rh2 | 343.883 g/mol

Used as a catalytic convertor in automobiles. It is also employed as an industrial catalyst in the manufacturing of chemical intermediates such as oxo-alcohols, as well as nitric acid and ethanoic (acetic) acid productions. It is, likewise, of interest for new capacitor electrode materials, dynamic random access memories (DRAMs) and non-volatile random access memories (FeRAMs).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39373-27-8
Molecular Formula H10O8Rh2
Molecular Weight (g/mol) 343.883
MDL Number MFCD00149843
InChI Key XZIIPNYQAMUKBU-UHFFFAOYSA-N
Synonym rhodium iii oxide pentahydrate, o8rh2, rhodium-platinum oxide, rhodium iii oxide 5-hydrate, rhodium iii oxide pentahydrate, premion
PubChem CID 45073257
IUPAC Name oxo(oxorhodiooxy)rhodium;pentahydrate
SMILES O.O.O.O.O.O=[Rh]O[Rh]=O

Specifications

Melting Point (decomposition)
Physical Form Powder
Quantity 0.25 g
Solubility Information Insoluble in water and aqua regia.
Formula Weight 343.88 (253.81 Anhydrous)
Percent Purity 99.99%
Grade Premion
Concentration or Composition (by Analyte or Components) Rh 59% min.
Assay (metals basis)
Chemical Name or Material Rhodium(III) oxide pentahydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.