missing translation for 'onlineSavingsMsg'
Learn More

Rhodium(III) oxide, anhydrous, 99.9% (metals basis), Rh 80.6% min

Product Code. 11341858
Change view
Click to view available options
Quantity:
0.5 g
2 g
Unit Size:
0.5g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11341858 0.5 g 0.5g
11351858 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11341858 Supplier Thermo Scientific Alfa Aesar Supplier No. 011814.02
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12036-35-0

Use in the catalytic converters of automobiles. It is also used as an industrial catalyst in the manufacturing of chemical intermediates such as oxo-alcohols, as well as nitric acid and ethanoic (acetic) acid production. It is also an area of interest for new capacitor electrode material researchers, dynamic random access memory (DRAMs) and non volatile random access memories (FeRAMs) makers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12036-35-0
MDL Number MFCD00011205

Specifications

Melting Point 1100°C (decomposition)
Color Black to Gray
Physical Form Powder
Quantity 0.5 g
Solubility Information Insoluble in water,acetone,conc. Hydrochloric acid,Nitric acid,Aqua regia.
Formula Weight 253.81
Percent Purity 99.9%
Concentration or Composition (by Analyte or Components) Rh 80.6% min.
Odor Odorless
Sensitivity Hygroscopic
Density 8.20 g/mL
Assay (metals basis)
Chemical Name or Material Rhodium(III) oxide, Anhydrous
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.