missing translation for 'onlineSavingsMsg'
Learn More

Rhodium(III) nitrate, solution, ca. 10% (w/v) Rh in 20-25 wt% HNO3

Product Code. 10502861 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
2 mL
10 mL
Packaging:
Glass bottle
Unit Size:
10mL
2mL
This item is not returnable. View return policy

Product Code. 10502861

Brand: Thermo Scientific Acros 318980020

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 10139-58-9 | N3O9Rh | 288.92 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10139-58-9, 7697-37-2, 7732-18-5
Molecular Formula N3O9Rh
Molecular Weight (g/mol) 288.92
MDL Number MFCD00011202
InChI Key VXNYVYJABGOSBX-UHFFFAOYSA-N
Synonym rhodium nitrate, rhodium iii nitrate, rhodium trinitrate, nitric acid, rhodium 3+ salt, rhodium 3+ trinitrate, nitric acid, rhodium 3+ salt 3:1, nitric acid,rhodium 3+ salt 3:1, rhodiumtrinitrate, rhodium +3 nitrate, acmc-20ajsk
PubChem CID 150190
IUPAC Name rhodium(3+);trinitrate
SMILES [Rh+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Color Brown-Orange to Brown-Red
Physical Form Solution
Quantity 2 mL
Linear Formula Rh(NO3)3
Formula Weight 288.92
Percent Purity 20 to 25% (HNO3) (w/w), 10 to 14% (Rh) (w/v)
Packaging Glass bottle
Specific Gravity 1.29
Density 1.2900g/mL
Chemical Name or Material Rhodium(III) nitrate, Solution, ca.10% (w/w) Rh in 20-25 weight% HNO3

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.