missing translation for 'onlineSavingsMsg'
Learn More

Rhodium(III) chloride solution, 99.5% (metals basis), Rh 10-30% w/w (cont. Rh)

Product Code. 15495015
Change view
Click to view available options
Quantity:
0.5 g
2 g
Unit Size:
0.5g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15495015 0.5 g 0.5g
15407245 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15495015 Supplier Thermo Scientific Alfa Aesar Supplier No. 046523.NJ
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 10049-07-7 | Cl3Rh | 209.26 g/mol

It is used as primary and secondary intermediate, in the preparation of compounds used in homogeneous catalysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10049-07-7
Molecular Formula Cl3Rh
Molecular Weight (g/mol) 209.26
MDL Number MFCD00011204
InChI Key SONJTKJMTWTJCT-UHFFFAOYSA-K
IUPAC Name rhodium(3+) trichloride
SMILES [Cl-].[Cl-].[Cl-].[Rh+3]

Specifications

Color Brown-Red
Linear Formula RhCl3
UN Number UN3264
Concentration Rh 10% w/w (cont. Rh)
Merck Index 14,8188
Solubility Information Immiscible in water.
Formula Weight 209.26
Percent Purity 99.5%
Quantity 0.5 g
Physical Form Liquid
Assay (metals basis)
Chemical Name or Material Rhodium (III) chloride solution
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.