missing translation for 'onlineSavingsMsg'
Learn More

Rhodium(III) chloride hydrate, Rh 38.0-45.5%

Product Code. 11365819
Change view
Click to view available options
Quantity:
0.25 g
1 g
5 g
25 g
Unit Size:
0.25g
1g
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11365819 25 g 25g
11341658 0.25 g 0.25g
11331658 1 g 1g
11351658 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11365819 Supplier Thermo Scientific Alfa Aesar Supplier No. 011032.14
 Restricted Item
Request Bulk or Custom Formats

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 20765-98-4 | Cl3Rh | 209.26 g/mol

Rhodium(III) chloride hydrate is used as a catalyst to prepare acetic acid. It is also used as a catalyst for reduction, oxidation of alkenes, hydration of acetylene and isomerization of alkenes. It is involved in the hydrosilylation of alfa,beta-unsaturated esters to form dimethylketene trimethylsilyl acetals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20765-98-4
Molecular Formula Cl3Rh
Molecular Weight (g/mol) 209.26
MDL Number MFCD00149839
InChI Key SONJTKJMTWTJCT-UHFFFAOYSA-K
Synonym Rhodium trichloride hydrate
IUPAC Name rhodium(3+) trichloride
SMILES [Cl-].[Cl-].[Cl-].[Rh+3]

Specifications

Melting Point 100°C (decomposition)
Color Red
Physical Form Crystalline
Quantity 25 g
Sensitivity Hygroscopic
Appearance Dark red crystals
Assay Percent Range 38.5 to 45.5%
Solubility Information Soluble in alcohol and water. Slightly soluble in acetone.
Formula Weight 209.26 (anhydrous)
Chemical Name or Material Rhodium(III) chloride hydrate, Rh 38.5-45.5%

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.