missing translation for 'onlineSavingsMsg'
Learn More

Rhodium(III) chloride hydrate, Premion™, 99.99% (metals basis)

Product Code. p-7058946
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11320298

Brand: Thermo Scientific Alfa Aesar 043702.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 20765-98-4 | Cl3Rh | 209.26 g/mol

Rhodium(III) chloride hydrate is used as a catalyst for the conversion of methane to acetic acid, isomerization of alkenes, reduction of aromatic rings and hydration of acetylene. It is involved in the hydrosilylation of alfa,beta-unsaturated esters to form dimethylketene trimethylsilyl acetals. It plays an important role as a catalyst for conjugate reduction of cinnamaldehydes followed by cross-coupling with arylboronic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20765-98-4
Molecular Formula Cl3Rh
Molecular Weight (g/mol) 209.26
MDL Number MFCD00149839
InChI Key SONJTKJMTWTJCT-UHFFFAOYSA-K
IUPAC Name rhodium(3+) trichloride
SMILES [Cl-].[Cl-].[Cl-].[Rh+3]

Specifications

Melting Point 100°C (decomposition)
Color Red
Physical Form Crystalline
Quantity 1 g
Merck Index 14,8188
Solubility Information Soluble in water and alcohol. Slightly soluble in acetone.
Formula Weight 209.26 (Anhydrous)
Percent Purity 99.99%
Grade Premion
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Rhodium (III) chloride hydrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.