missing translation for 'onlineSavingsMsg'
Learn More

Rhodium(III) chloride, anhydrous, 99.9% (metals basis), Rh 48.7% min, Thermo Scientific Chemicals

Catalog Number 11361858
Click to view available options
Quantity:
0.25 g
1 g
5 g
10049-07-7
Cl3Rh
209.26
MFCD00011204
SONJTKJMTWTJCT-UHFFFAOYSA-K
rhodium(3+) trichloride
[Cl-].[Cl-].[Cl-].[Rh+3]
10049-07-7
Cl3Rh
209.26
MFCD00011204
SONJTKJMTWTJCT-UHFFFAOYSA-K
rhodium(3+) trichloride
[Cl-].[Cl-].[Cl-].[Rh+3]

It is used in homogeneous catalysis, for example in the industrial prodcution of acetic acid. It is used to coat optic fibres and optical mirrors, and for crucibles, thermocouple elements and headlight reflectors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Red to Brown
Physical Form Powder
Quantity 0.25 g
Linear Formula RhCl3
Merck Index 14,8188
Solubility Information Insoluble in water. Soluble in hydroxide and cyanide solutions.
Formula Weight 209.26
Percent Purity 99.9%
Concentration or Composition (by Analyte or Components) Rh 48.7% min.
Odor Odorless
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Rhodium(III) chloride, Anhydrous
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'